Search Thermo Fisher Scientific
FIGURE: 1 / 3
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt.
Reconstitute by adding 50 uL deionized water.
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.