Search Thermo Fisher Scientific
The PA1-31194 immunogen is: Synthetic peptide unique to PAC9 protein was used to generate antibodies in rabbit. (unfortunately, the amino acid sequence is considered to be commercially sensitive). All peptides were amidated before conjugation.
Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. It is regulated by a family of G protein-coupled receptors, protein kinases, and calcium. The type 9 adenylyl cyclase is a widely distributed adenylyl cyclase, and it is stimulated by beta-adrenergic receptor activation but is insensitive to forskolin, calcium, and somatostatin.