Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total CI098.
Enables AMP binding activity and nucleobase-containing compound kinase activity. Involved in nucleoside diphosphate phosphorylation and nucleoside triphosphate biosynthetic process. Located in 9+2 motile cilium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate kinase 8; AK 8; ATP-AMP transphosphorylase 8; HypB, AroK, ADK and rho factor domain containing protein RGD1303144; putative adenylate kinase-like protein C9orf98; putative adenylate kinase-like protein C9orf98 homolog; RP11-143F18.1
Gene Aliases: AK 8; AK8; C9orf98; DDX31; RGD1303144
UniProt ID: (Human) Q96MA6, (Rat) Q68FP8
Entrez Gene ID: (Human) 158067, (Rat) 311833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support