Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CHSRSADVDS LSESRTQESA CLELDPAAQS LASLAPIGAQ WP
Adropin is involved in the regulation of glucose homestasis and lipid metabolism. It is a secreted protein encoded by the Energy Homeostasis Associated gene (ENHO) found in the brain and liver. In the liver, the protein has been found to increase in response to high-fat foods and decrease after fasting. The protein seems to play a role in controlling the activity of other metabolic genes, specifically those involved in the production of lipids from carbohydrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adropin; Energy homeostasis-associated protein; GAAI470
Gene Aliases: C9orf165; ENHO; UNQ470; UNQ470/PRO830
UniProt ID: (Human) Q6UWT2
Entrez Gene ID: (Human) 375704
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support