Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Contact Us
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Aggrecan Neo Antibodies

            Invitrogen

            Aggrecan Neo Polyclonal Antibody

            Advanced Verification
            17 Published Figures
            25 References

            Promotions

            View available promotion(s)

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # PA1-1746
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (MXN)
            11,934.49
            100 µg
             
            Check your price & availability
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite Aggrecan Neo Polyclonal Antibody

            • Antibody Testing Data (1)
            • Published Figures (17)
            • Advanced Verification (1)
            Application
            Aggrecan Neo Antibody in Immunohistochemistry (IHC)
            Group 53 Created with Sketch.

            FIGURE: 1 / 19

            Aggrecan Neo Antibody (PA1-1746) in IHC

            Immunofluorescent analysis of Aggrecan Neo in OA cartilage using a polyclonal antibody (Product # PA1-1746).
            View Product

            Aggrecan Neo Antibody in Immunohistochemistry (IHC)
            Aggrecan Neo Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            Aggrecan Neo Antibody in Immunocytochemistry (ICC/IF)
            Aggrecan Neo Antibody in Immunocytochemistry (ICC/IF)
            Aggrecan Neo Antibody in Immunocytochemistry (ICC/IF)
            Aggrecan Neo Antibody in Immunocytochemistry (ICC/IF)
            Aggrecan Neo Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Aggrecan Neo Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Aggrecan Neo Antibody in Immunohistochemistry (IHC)
            Aggrecan Neo Antibody in Western Blot, Immunohistochemistry (WB, IHC)
            Aggrecan Neo Antibody in Immunohistochemistry (IHC)
            Aggrecan Neo Antibody in Immunohistochemistry (IHC)
            Aggrecan Neo Antibody in Western Blot (WB)
            Aggrecan Neo Antibody in Western Blot (WB)
            Aggrecan Neo Antibody in Western Blot (WB)
            Aggrecan Neo Antibody in Western Blot (WB)
            Aggrecan Neo Antibody in Western Blot (WB)
            Aggrecan Neo Antibody in Western Blot (WB)
            Aggrecan Neo Antibody

            Product Details

            PA1-1746

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1 µg/mL
            View 11 publications 11 publications

            Immunohistochemistry (IHC)

            Assay-dependent
            View 8 publications 8 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            -
            View 3 publications 3 publications

            Immunocytochemistry (ICC/IF)

            -
            View 3 publications 3 publications
            Product Specifications

            Species Reactivity

            Bovine, Dog, Human, Mouse, Pig, Rat

            Published species

            Bovine, Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Synthetic peptide corresponding to residues C G G N(387) I T E G E(392) of human aggrecan.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.83 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS with 1mg/mL BSA

            Contains

            0.05% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2242021

            Product Specific Information

            PA1-1746 detects human, rat, mouse, bovine, canine, and porcine aggrecan neo G1.

            PA1-1746 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~64 kDa protein representing the neoepitope sequence (NITEGE) generated by aggrecanase-mediated cleavage at Glu373-Ala374 of aggrecan core proteins.

            The PA1-1746 immunogen is a synthetic peptide corresponding to residues residues C G G N(387) I T E G E(392) of human aggrecan.

            Target Information

            Aggrecan also known as cartilage-specific proteoglycan core protein (CSPCP) or chondroitin sulfate proteoglycan 1 is a protein that is encoded by the ACAN gene. The encoded protein is an integral part of the extracellular matrix in cartilaginous tissue and it withstands compression in cartilage. The human form of the protein is 2316 amino acids long (approximately 250 kDa) and can be expressed in multiple isoforms due to alternative splicing. Aggrecan exhibits a bottlebrush structure, in which chondroitin sulfate and keratan sulfate chains are attached to an extended protein core. The synthesis and degradation of aggrecan are being investigated for their roles in cartilage deterioration during joint injury, disease, and aging. It contains three interglobular domains (IGDs), G1, G2, and G3 that are involved in aggregation, hyaluronan binding, cell adhesion, and chondrocyte apoptosis. Aggrecan fragments from articular cartilage are released into the synovial fluid at all stages of human osteoarthritis as a result of cleavage by aggrecanase and MMP activity. Antibodies BC-3 and BC-4 can be used to detect these neo epitopes of Aggrecanase (ARGSVXXX) and MMP (XXXVDIPEN), respectively.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Bioinformatics

            Protein Aliases: aggrecan 1; aggrecan 1 (chondroitin sulfate proteoglycan 1, large aggregating proteoglycan, antigen identified by monoclonal antibody A0122); Aggrecan core protein; aggrecan epidermal growth factor-like domain-1; aggrecan, structural proteoglycan of cartilage; cartilage aggregating proteoglycan; Cartilage-specific proteoglycan core protein; Chondroitin sulfate proteoglycan 1; Chondroitin sulfate proteoglycan core protein 1; CSPCP; high density cartilage proteoglycan; large aggregating proteoglycan; large aggregating proteoglycan, antigen

            View more View less

            Gene Aliases: ACAN; Agc; AGC1; AGCAN; b2b183Clo; cmd; CSPCP; CSPG1; CSPGCP; MSK16; SEDK

            View more View less

            UniProt ID: (Bovine) P13608, (Human) Q9UCP4, (Rat) P07897, (Mouse) Q61282

            View more View less

            Entrez Gene ID: (Dog) 403828, (Pig) 397255, (Bovine) 280985, (Human) 176, (Rat) 58968, (Mouse) 11595

            View more View less

            Function(s)

            calcium ion binding hyaluronic acid binding carbohydrate binding protein binding extracellular matrix structural constituent metal ion binding extracellular matrix glycoprotein

            Process(es)

            cell adhesion skeletal system development proteolysis central nervous system development keratan sulfate biosynthetic process extracellular matrix disassembly extracellular matrix organization keratan sulfate catabolic process cartilage condensation ossification chondrocyte development response to radiation response to mechanical stimulus response to gravity response to glucose multicellular organism aging response to acidic pH response to organic cyclic compound spinal cord development proteoglycan biosynthetic process collagen fibril organization negative regulation of cell migration response to drug chondroblast differentiation cellular response to growth factor stimulus
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            méxico flag icon
            México

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-p2f5s:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0