Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Predicted species reactivity based on immunogen sequence: Donkey: (85%), Horse: (85%), Pig: (85%), Rabbit: (85%), Chicken: (80%)
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Albumin; albumin (32 AA); albumin (AA 34); albumin 1; cell growth inhibiting protein 42; growth-inhibiting protein 20; serum albumin
Gene Aliases: ALB; Alb-1; Alb1; Albza; ANALBA; FDAH; GIG20; GIG42; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; UNQ696/PRO1341
UniProt ID: (Human) Q8IUK7, (Rat) P02770, (Mouse) P07724
Entrez Gene ID: (Human) 213, (Rat) 24186, (Mouse) 11657
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support