Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPHSYPALS AEQKKELSDI ALRIVAPGKG ILAADESVGS MAKRLSQIGV ENTEENRRLY RQVLFSADDR VKKCIGGVIF FHETLYQKDD NGVPFVRTIQ DKGIVVGIKV DKGVVPLAGT DGETTTQGLD GLSERCAQYK KDGADFAKWR CVLKISERTP SALAILENAN VLARYASICQ QNGIVPIVEP EILPDGDHDL KRCQYVTEKV LAAVYKALSD HHVYLEGTLL KPNMVTPGHA CPIKYTPEEI AMATVTALRR TVPPAVPGVT FLSGGQSEEE ASFNLNAINR CPLPRPWALT FSYGRALQAS ALNAWRGQRD NAGAATEEFI KRAEVNGLAA QGKYEGSGED GGAAAQSLYI ANHAY (1-364 aa encoded by BC003613)
This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq, Jul 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aldolase 3; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; Brain-type aldolase; EC 4.1.2.13; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; Fructose-bisphosphate aldolase C; Scrapie-responsive protein 2; Zebrin II
Gene Aliases: AI847350; ALDC; ALDCAA; Aldo3; ALDOC; AU040929; F16dip7; RATALDCAA; Scrg2
UniProt ID: (Human) P09972, (Mouse) P05063, (Rat) P09117
Entrez Gene ID: (Human) 230, (Mouse) 11676, (Rat) 24191
Molecular Function:
aldolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support