Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Promo Code: RACCS25 Save on advanced culture solutions Save up to 22% on optimized formulations to streamline your neural, stem and cancer cell culture. Learn more
Immunogen sequence: MNGDSRAAV VTSPPPTTAP HKERYFDRVD ENNPEYLRER NMAPDLRQDF NMMEQKKRVS MILQSPAFCE ELESMIQEQF KKGKNPTGLL ALQQIADFMT TNVPNVYPAA PQGGMAALNM SLGMVTPVND LRGSDSIAYD KGEKLLRCKL AAFYRLADLF GWSQLIYNHI TTRVNSEQEH FLIVPFGLLY SEVTASSLVK INLQGDIVDR GSTNLGVNQA GFTLHSAIYA ARPDVKCVVH IHTPAGAAVS AMKCGLLPIS PEALSLGEVA YHDYHGILVD EEEKVLIQKN LGPKSKVLIL RNHGLVSVGE SVEEAFYYIH NLVVACEIQV RTLASAGGPD NLVLLNPEKY KAKSRSPGSP VGEGTGSPPK WQIGEQEFEA LMRMLDNLGY RTSSSGRRQD G (1-400 aa encoded by BC013393)
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADD2; ADDB; adducin 1 (alpha); Adducin beta; Adducin-1; Adducin-alpha; Adducin-beta; Adducin1; Alpha adducin; alpha-ADD; Alpha-adducin; beta Adducin; Beta-adducin; erythrocyte adducin alpha subunit; Erythrocyte adducin subunit alpha
Gene Aliases: ADD1; ADDA; AI256389
UniProt ID: (Human) P35611, (Mouse) Q9QYC0, (Rat) Q63028
Entrez Gene ID: (Human) 118, (Mouse) 11518, (Rat) 24170
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support