Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alsin; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) homolog; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; Amyotrophic lateral sclerosis 2 protein homolog; amyotrophic lateral sclerosis protein 2 homolog; FLJ31851; MGC87187
Gene Aliases: 3222402C23Rik; 9430073A21Rik; ALS2; ALS2CR6; Alsin; ALSJ; IAHSP; KIAA1563; mKIAA1563; PLSJ
UniProt ID: (Human) Q96Q42, (Mouse) Q920R0
Entrez Gene ID: (Human) 57679, (Mouse) 74018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support