Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 79%; Dog: 86%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Sheep: 79%
This gene encodes the nonamelogenin enamel matrix protein ameloblastin. The encoded protein may be important in enamel matrix formation and mineralization. This gene is located in the calcium-binding phosphoprotein gene cluster on chromosome 4. Mutations in this gene may be associated with dentinogenesis imperfect and autosomal dominant amylogenesis imperfect.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ameloblastin; ameloblastin (enamel matrix protein); enamel matrix protein
Gene Aliases: AI1F; AMBN
UniProt ID: (Human) Q9NP70
Entrez Gene ID: (Human) 258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support