Save up to 31% on select cell culture essentials

Bundle and save

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Custom Products & Projects
              • Instrument Management

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • Angiopoietin 1 Antibodies

            Invitrogen

            Angiopoietin 1 Polyclonal Antibody

            View all (29) Angiopoietin 1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # PA5-46968
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (TWD)
            20,080.00
            Online offer
            Ends: 30-Jun-2025
            25,600.00
            Save 5,520.00 (22%)
            100 µg
             
            Check your price & availability
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite Angiopoietin 1 Polyclonal Antibody

            • Antibody Testing Data (3)
            Application
            Angiopoietin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            Angiopoietin 1 Antibody (PA5-46968) in IHC (P)

            Immunohistochemical analysis of Angiopoietin 1 in immersion fixed paraffin-embedded sections of human prostate array. Samples were incubated in Angiopoietin 1 polyclonal antibody (Product # PA5-46968) using a dilution of 5 µg/mL overnight at 4 °C. Tissue was stained using the Anti-Goat HRP-DAB Cell & Tissue Staining Kit (brown) and counterstained with hematoxylin (blue). Lower panel shows a lack of labeling... View More
            View Product

            Angiopoietin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Angiopoietin 1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Angiopoietin 1 Antibody in Immunohistochemistry (Frozen) (IHC (F))

            Product Details

            PA5-46968

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            0.1 µg/mL
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            5-15 µg/mL
            -

            Immunohistochemistry (Frozen) (IHC (F))

            5-15 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human, Mouse

            Host/Isotype

            Goat / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-1 Ser20-Phe498
            View immunogen

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Concentration

            0.2 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS with 5% trehalose

            Contains

            No Preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2607762

            Product Specific Information

            In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) Angiopoietin-2, rhAngiopoietin-4, and rhAngiopoietin-like Factor/CDT6 is observed.

            Reconstitute at 0.2 mg/mL in sterile PBS.

            Target Information

            Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: ANG-1; Angiopoietin-1

            View more View less

            Gene Aliases: 1110046O21Rik; AGP1; AGPT; Ang-1; ANG1; ANGPT1; KIAA0003

            View more View less

            UniProt ID: (Human) Q15389, (Mouse) O08538

            View more View less

            Entrez Gene ID: (Human) 284, (Mouse) 11600

            View more View less

            Function(s)

            Ras guanyl-nucleotide exchange factor activity receptor tyrosine kinase binding receptor binding vascular endothelial growth factor receptor binding intercellular signal molecule

            Process(es)

            MAPK cascade in utero embryonic development negative regulation of protein phosphorylation regulation of endothelial cell proliferation sprouting angiogenesis positive regulation of receptor internalization negative regulation of cytokine secretion involved in immune response negative regulation of cell adhesion activation of transmembrane receptor protein tyrosine kinase activity positive regulation of endothelial cell migration positive regulation of phosphatidylinositol 3-kinase signaling regulation of skeletal muscle satellite cell proliferation hemopoiesis cell differentiation heparin biosynthetic process positive regulation of protein ubiquitination cell-substrate adhesion regulation of tumor necrosis factor production positive regulation of peptidyl-serine phosphorylation protein localization to cell surface negative regulation of protein import into nucleus negative regulation of apoptotic process negative regulation of vascular permeability regulation of I-kappaB kinase/NF-kappaB signaling regulation of protein binding negative regulation of neuron apoptotic process positive regulation of blood vessel endothelial cell migration positive regulation of GTPase activity positive regulation of cell adhesion Tie signaling pathway positive regulation of peptidyl-tyrosine phosphorylation leukocyte migration positive chemotaxis positive regulation of protein kinase B signaling positive regulation of ERK1 and ERK2 cascade glomerulus vasculature development negative regulation of endothelial cell apoptotic process regulation of macrophage migration inhibitory factor signaling pathway angiogenesis ovarian follicle development patterning of blood vessels vasculogenesis endocardium morphogenesis transmembrane receptor protein tyrosine kinase signaling pathway multicellular organism development endoderm development positive regulation of vascular endothelial growth factor receptor signaling pathway protein homooligomerization cardiac muscle tissue morphogenesis vasculogenesis involved in coronary vascular morphogenesis
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Taiwan flag icon
            Taiwan

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0