Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Ankyrin-G is found in several tissues including brain where, due to tissue-specific RNA processing, it is expressed as two different isoforms, localized to axon initial segments and nodes of Ranvier. Ankyrin-G is also found at sites of epithelial cell-cell contact and in the intracellular membranes of some cells. Ankyrin-G isoforms contain a unique stretch of sequence highly enriched in serine and threonine residues immediately following a globular head domain. Ankyrin-G is thought to participate in the maintenance and targeting of ion channels and cell adhesion molecules to the nodes of Ranvier and to axon initial segments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANK-3; ankyrin 3 (G); ankyrin 3, epithelial; Ankyrin-3; Ankyrin-G; brain-specific ankyrin-G
Gene Aliases: 2900054D09Rik; AI314020; ANK-3; ANK3; AnkG; Ankyrin-3; ANKYRIN-G; MRT37
UniProt ID: (Human) B4DIL1, (Rat) O70511, (Mouse) G5E8K5
Entrez Gene ID: (Human) 288, (Rat) 361833, (Mouse) 11735
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support