Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Human cervix carcinoma
Cellular location: Membrane.
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Annexin A7; Annexin VII; Annexin-7; Interleukin-8; RP11-537A6.8; Synexin
Gene Aliases: ANX7; ANXA7; OK/SW-cl.95; SNX; SYNEXIN
UniProt ID: (Human) P20073
Entrez Gene ID: (Human) 310
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support