Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAQVLRGTV TDFPGFDERA DAETLRKAMK GLGTDEESIL TLLTSRSNAQ RQEISAAFKT LFGRDLLDDL KSELTGKFEK LIVALMKPSR LYDAYELKHA LKGAGTNEKV LTEIIASRTP EELRAIKQVY EEEYGSSLED DVVGDTSGYY QRMLVVLLQA NRDPDAGIDE AQVEQDAQAL FQAGELKWGT DEEKFITIFG TRSVSHLRKV FDKYMTISGF QIEETIDRET SGNLEQLLLA VVKSIRSIPA YLAETLYYAM KGAGTDDHTL IRVMVSRSEI DLFNIRKEFR KNFATSLYSM IKGDTSGDYK KALLLLCGED D (1-320 aa encoded by BC001429)
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anchorin CII; Annexin A5; Annexin V; Annexin-5; Calphobindin I; CBP-I; CPB-I; Endonexin II; epididymis secretory protein Li 7; Lipocortin V; PAP-I; Placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha
Gene Aliases: ANX5; ANXA5; ENX2; HEL-S-7; PP4; R74653; RPRGL3
UniProt ID: (Human) P08758, (Mouse) P48036
Entrez Gene ID: (Human) 308, (Mouse) 11747
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support