Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description: BMS147 specifically recognizes Annexin V (VAC- alpha ). BMS147 is suitable for ELISA application, Western blot analysis and is suitable as primary antibody for staining in FACS analysis.
BMS147FI can be used for direct staining of cells.
BMS147BT can be used in a two-step procedure using the biotin-(strept-)avidin system. For intracellular stainings the use of BMS147FI is recommended.
Applications Tested: ELISA, Flow Cytometry (Intracellular Staining), Western Blotting.
Purity: >95%.
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anchorin CII; Annexin A5; Annexin V; Annexin-5; Calphobindin I; CBP-I; CPB-I; Endonexin II; epididymis secretory protein Li 7; Lipocortin V; PAP-I; Placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha
Gene Aliases: ANX5; ANXA5; ENX2; HEL-S-7; PP4; RPRGL3
UniProt ID: (Human) P08758
Entrez Gene ID: (Human) 308
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support