Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AKVLLHIAT EDNIGGEEGN MKFGTTLKNC RHLLECAKEL DVQIIGVKFH VSSACKESQV YVHALSDARC VFDMAGEIGF TMNMLDIGGG FTGTEFQLEE VNHVISPLLD IYFPEGSGVK IISEPGSYYV SSAFTLAVNI IAKKVVENDK FPSGVEKTGS DEPAFMYYMN DGVYGSFASK LSEDLNTIPE VHKKYKEDEP LFTSSLWGPS CDELDQIVES CLLPELNVGD WLIFDNMGAD SFHEPSAFND FQRPAIYYMM SFSDWYEMQD AGITSDSMMK NFFFVPSCIQ LSQEDSFSAE A (149-448 aa encoded by BC019279)
Antizyme inhibitor (AZIN1) protein that positively regulates ornithine decarboxylase (ODC) activity and polyamine uptake. AZI is an enzymatically inactive ODC homolog that counteracts the negative effect of ODC antizymes (AZs) OAZ1, OAZ2 and OAZ3 on ODC activity by competing with ODC for antizyme-binding. Inhibits antizyme-dependent ODC degradation and releases ODC monomers from their inactive complex with antizymes, leading to formation of the catalytically active ODC homodimer and restoring polyamine production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antizyme inhibitor 1; AZI; O OAZIN; ODC antizyme inhibitor; Ornithine decarboxylase antizyme inhibitor
Gene Aliases: 1700085L02Rik; AZI; AZIA1; AZIN1; OAZI; OAZIN; ODC1L
UniProt ID: (Human) O14977, (Mouse) O35484
Entrez Gene ID: (Human) 51582, (Rat) 58961, (Mouse) 54375
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support