Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alternative name: G3a, NG20; Apo M; Apo-M; Apolipoprotein M; MGC22400; NG20 like protein; NG20-like protein; Protein G3a; Protein Px
Gene Aliases: 1190010O19Rik; apo-M; APOM; G3A; HSPC336; NG20
UniProt ID: (Human) O95445, (Mouse) Q9Z1R3, (Rat) P14630
Entrez Gene ID: (Human) 55937, (Mouse) 55938, (Rat) 55939
Molecular Function: apolipoprotein transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support