Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Proteintech
This antibody was generated against the C-terminal region of human ApoA1.
Immunogen sequence: RHFWQQDEP PQSPWDRVKD LATVYVDVLK DSGRDYVSQF EGSALGKQLN LKLLDNWDSV TSTFSKLREQ LGPVTQEFWD NLEKETEGLR QEMSKDLEEV KAKVQPYLDD FQKKWQEEME LYRQKVEPLR AELQEGARQK LHELQEKLSP LGEEMRDRAR AHVDALRTHL APYSDELRQR LAARLEALKE NGGARLAEYH AKATEHLSTL SEKAKPALED LRQGLLPVLE SFKVSFLSAL EEYTKKLNTQ (19-267 aa encoded by BC005380)
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-Sep; 2-Sep; Apo a; apo A-1; APO A1; APO I; Apo-AI; ApoAI; Apolipoprotein; Apolipoprotein A-I; Apolipoprotein A1; Apolipoprotein AI; MGC102525; MGC117399; MGC124929; Udpgt; Ugt1; Ugt1a7
Gene Aliases: Alp-1; apo(a); apo-AI; Apoa-1; apoA-I; APOA1; Brp-14; Ltw-1; Lvtw-1; Sep-1; Sep-2; Sep2
UniProt ID: (Human) P02647, (Mouse) Q00623
Entrez Gene ID: (Human) 335, (Mouse) 11806
Molecular Function: apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support