Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive control: Human lung tissue.
This gene encodes a member of the histidine triad (HIT) superfamily, some of which have nucleotide-binding and diadenosine polyphosphate hydrolase activities. The encoded protein may play a role in single-stranded DNA repair. Mutations in this gene have been associated with ataxia-ocular apraxia. Multiple transcript variants encoding distinct isoforms have been identified for this gene, however, the full length nature of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aprataxin; FHA-HIT; Forkhead-associated domain histidine triad-like protein
Gene Aliases: 2410016G21Rik; AA388047; AOA; AOA1; APTX; AXA1; EAOH; EOAHA; FHA-HIT
UniProt ID: (Human) Q7Z2E3, (Rat) Q8K4H4, (Mouse) Q7TQC5
Entrez Gene ID: (Human) 54840, (Rat) 259271, (Mouse) 66408
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support