Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AQP3 is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 3 is localized at the basal lateral membranes of collecting duct cells in the kidney. In addition to its water channel function, aquaporin 3 has been found to facilitate the transport of nonionic small solutes such as urea and glycerol, but to a smaller degree. It has been suggested that water channels can be functionally heterogeneous and possess water and solute permeation mechanisms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 31.4 kDa water channel protein; AQP 3; AQP-3; Aquaglyceroporin-3; aquaporin 3 (GIL blood group); aquaporin 3 (Gill blood group); Aquaporin-3; Aquaporin3; Gill blood group
Gene Aliases: AQP-2; AQP-3; AQP3; GIL
UniProt ID: (Human) Q92482, (Bovine) Q08DE6, (Rat) P47862, (Mouse) Q8R2N1
Entrez Gene ID: (Human) 360, (Sheep) 443047, (Bovine) 780866, (Rat) 65133, (Mouse) 11828
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support