Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 5
Antibody detects endogenous levels of total AQP6.
Aquaporin 6 forms a water-specific channel that participates in distinct physiological functions such as glomerular filtration, tubular endocytosis and acid-base metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AQP-6; aquaporin 2-like, kidney specific; aquaporin 6, kidney specific; Aquaporin-2-like; Aquaporin-6; aquaporin-6, kidney specific; hKID; Kidney-specific aquaporin
Gene Aliases: AI790337; AQP2L; AQP6; KID
UniProt ID: (Human) Q13520, (Rat) Q9WTY0, (Mouse) Q8C4A0
Entrez Gene ID: (Human) 363, (Rat) 29170, (Mouse) 11831
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support