Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AQP-7; AQPap; Aquaglyceroporin-7; Aquaporin adipose; Aquaporin-7; Aquaporin-7-like; MGC149555; MGC149556; RP11-311H10.6
Gene Aliases: AQP7; AQP7L; AQP9; AQPap; GLYCQTL
UniProt ID: (Human) O14520, (Rat) P56403, (Mouse) O54794
Entrez Gene ID: (Human) 364, (Bovine) 615498, (Sheep) 101103697, (Rat) 29171, (Mouse) 11832
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support