Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This antibody detects endogenous protein at a molecular weight of 40 kDa.
Purity is >95% by SDS-PAGE.
ARFIP1 gene ontology annotations related to this gene include regulation of protein secretion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor-interacting protein 1; arfaptin 1; Arfaptin-1; Arfaptin1; ARFP1
Gene Aliases: 2310030I10Rik; Arfaptin; ARFIP1; D3Ertd598e; HSU52521
UniProt ID: (Human) P53367, (Rat) Q9JHU5, (Mouse) G5E8V9
Entrez Gene ID: (Human) 27236, (Rat) 60382, (Mouse) 99889
Molecular Function: vesicle coat protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support