Product References
Development and Characterization of Inducible Astrocyte-Specific Aromatase Knockout Mice.
Biology
Wang J,Pratap UP,Lu Y,Sareddy GR,Tekmal RR,Vadlamudi RK,Brann DW
PA1-21398 was used in Western Blotting to develop a tamoxifen (TMX)-inducible astrocyte-specific aromatase knockout mouse line (GFAP-ARO-iKO mice) to specifically deplete the E2 synthesis enzymes and aromatase in astrocytes after their development in adult mice.
Wed Apr 19 00:00:00 UTC 2023
KISS1R and ANKRD31 Cooperate to Enhance Leydig Cell Gene Expression via the Cytoskeletal-Nucleoskeletal Pathway.
Frontiers in cell and developmental biology
Ricci G,Guillou F,Catizone A,Mele VG,Moggio M,Chioccarelli T,Diano N,Meccariello R,Pierantoni R,Fasano S,Cobellis G,Chianese R,Manfrevola F
PA1-21398 was used in Western Blotting to highlight a novel role for the Kisspeptin-10 in the induction of the cytoskeletal-nucleoskeletal route, downstream a physical interaction between KISS1 receptor and ANKRD31, with gene expression activation as final effect, in Leydig cells.
Sat Jul 16 00:00:00 UTC 2022
High Doses of D-Chiro-Inositol Alone Induce a PCO-Like Syndrome and Other Alterations in Mouse Ovaries.
International journal of molecular sciences
Bevilacqua A,Dragotto J,Lucarelli M,Di Emidio G,Monastra G,Tatone C
PA1-21398 was used in Western Blot to analyze effects of higher doses of DCIns, 5, 10 and 20 mg/day, administered to young adult female mice for 21 days, on ovarian histology, serum testosterone levels and expression of the ovarian enzyme aromatase.
Wed May 26 00:00:00 UTC 2021
High Doses of D-Chiro-Inositol Alone Induce a PCO-Like Syndrome and Other Alterations in Mouse Ovaries.
International journal of molecular sciences
Bevilacqua A,Dragotto J,Lucarelli M,Di Emidio G,Monastra G,Tatone C
PA1-21398 was used in Western Blot to analyze effects of higher doses of DCIns, 5, 10 and 20 mg/day, administered to young adult female mice for 21 days, on ovarian histology, serum testosterone levels and expression of the ovarian enzyme aromatase.
Wed May 26 00:00:00 UTC 2021
The role of vaspin in porcine corpus luteum.
The Journal of endocrinology
Kurowska P,Mlyczyńska E,Dawid M,Grzesiak M,Dupont J,Rak A
PA1-21398 was used in Western Blot to find that vaspin/GRP78 expression depends on the luteal phase stage and vaspin affects luteal cells endocrinology, indicating that vaspin is a new regulator of luteal cells steroidogenesis and CL formation.
Tue Dec 01 00:00:00 UTC 2020
Role of vaspin in porcine ovary: effect on signaling pathways and steroid synthesis via GRP78 receptor and protein kinase A†.
Biology of reproduction
Kurowska P,Mlyczyńska E,Dawid M,Dupont J,Rak A
Published figure using Aromatase polyclonal antibody (Product # PA1-21398) in Western Blot
Tue May 26 00:00:00 UTC 2020
INSIGHTS ON AROMATASE IMMUNOHISTOCHEMISTRY: VARIATIONS BETWEEN INTRINSIC MOLECULAR SUBTYPES OF BREAST CANCERS.
Acta endocrinologica (Bucharest, Romania : 2005)
Armasu I,Preda C,Ianole V,Mocanu V,Hristov I,Andriescu EC,Cretu-Silivestru I,Vasiliu I,Dascalu CG,Lupascu CD,Crumpei I,Serban DN,Serban IL,Ciobanu Apostol DG
PA1-21398 was used in Immunohistochemistry to describe aromatase expression in the major tissue compartments of breast malignant tumors.
Fri Apr 15 00:00:00 UTC 2022
Developmental programming: Changes in mediators of insulin sensitivity in prenatal bisphenol A-treated female sheep.
Reproductive toxicology (Elmsford, N.Y.)
Puttabyatappa M,Martin JD,Andriessen V,Stevenson M,Zeng L,Pennathur S,Padmanabhan V
PA1-21398 was used in Western Blotting to determine if changes in insulin sensitivity mediators (increase in inflammation, oxidative stress, and lipotoxicity and/or decrease in adiponectin) and the intracrine steroidal milieu contributes to these metabolic perturbations.
Mon Apr 01 00:00:00 UTC 2019
Tanshinone IIA attenuates estradiol-induced polycystic ovarian syndrome in mice by ameliorating FSHR expression in the ovary.
Experimental and therapeutic medicine
Jin J,Hu QY,Xu WW,Zhu WJ,Liu B,Liu J,Wang W,Zhou HF
PA1-21398 was used in Western Blotting to replicate PCOS via the neonatal treatment of estradiol in mice.
Wed May 01 00:00:00 UTC 2019
Increased expression of aromatase cytochrome P450 enzyme is associated with prolactinoma invasiveness in post-menopausal women.
The Journal of international medical research
Su YX,Du GL,Shen HL,Wang W,Bao JL,Aierken A,Wang BW,Jiang S,Zhu J,Gao XM
Published figure using Aromatase polyclonal antibody (Product # PA1-21398) in Immunohistochemistry
Mon Jul 01 00:00:00 UTC 2019
Developmental Programming: Impact of Prenatal Testosterone Excess on Steroidal Machinery and Cell Differentiation Markers in Visceral Adipocytes of Female Sheep.
Reproductive sciences (Thousand Oaks, Calif.)
Puttabyatappa M,Lu C,Martin JD,Chazenbalk G,Dumesic D,Padmanabhan V
PA1-21398 was used in Western Blotting to show that increased commitment of AT-MSCs to adipocyte lineage and decreased differentiation to adipocytes may underlie the small adipocyte phenotype of prenatal T-treated females.
Sun Jul 01 00:00:00 UTC 2018
Aromatase enzyme expression in acromegaly and its possible relationship with disease prognosis.
Endocrine
Selek A,Cetinarslan B,Gurbuz Y,Tarkun I,Canturk Z,Cabuk B
PA1-21398 was used in immunohistochemistry to assess the relationship between aromatase expression and the clinical and prognostic features growth hormone secreting adenomas, prolactinomas, and normal pituitary tissues
Fri May 01 00:00:00 UTC 2015
Developmental programming: prenatal steroid excess disrupts key members of intraovarian steroidogenic pathway in sheep.
Endocrinology
Padmanabhan V,Salvetti NR,Matiller V,Ortega HH
Published figure using Aromatase polyclonal antibody (Product # PA1-21398) in Immunohistochemistry
Mon Sep 01 00:00:00 UTC 2014
Developmental programming: prenatal steroid excess disrupts key members of intraovarian steroidogenic pathway in sheep.
Endocrinology
Padmanabhan V,Salvetti NR,Matiller V,Ortega HH
Published figure using Aromatase polyclonal antibody (Product # PA1-21398) in Immunohistochemistry
Mon Sep 01 00:00:00 UTC 2014
Pathogen-associated molecular patterns initiate inflammation and perturb the endocrine function of bovine granulosa cells from ovarian dominant follicles via TLR2 and TLR4 pathways.
Endocrinology
Price JC,Bromfield JJ,Sheldon IM
PA1-21398 was used in western blot to study the role of TLR2 and TLR4 in the initiation of inflammation and disturbance of bovine granulosa cell endocrine function by bacterial PAMPs
Sun Sep 01 00:00:00 UTC 2013