Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and catalyzes the last steps of estrogen biosynthesis, three successive hydroxylations of the A ring of androgens. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation. The gene expresses two transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aromatase; CYPXIX; Cytochrome P-450AROM; Cytochrome P450 19A1; cytochrome P450, family 19, subfamily A, polypeptide 1; cytochrome P450, subfamily XIX (aromatization of androgens); Estrogen synthase; estrogen synthetase; flavoprotein-linked monooxygenase; MGC104309; microsomal monooxygenase
Gene Aliases: ARO; ARO1; CPV1; CYAR; CYP19; CYP19A1; CYPXIX; P-450AROM
UniProt ID: (Human) P11511
Entrez Gene ID: (Human) 1588
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support