Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31604 has been tested in indirect ELISA.
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-like protein 3; Actin-related protein 3; ARP3 actin-related protein 3 homolog
Gene Aliases: 1200003A09Rik; ACTR3; ARP3
UniProt ID: (Human) P61158, (Mouse) Q99JY9
Entrez Gene ID: (Human) 10096, (Mouse) 74117
Molecular Function:
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support