Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein encoded by gene ARPC1A. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages. This protein also has a role in centrosomal homeostasis by being an activator and substrate of the Aurora A kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin related protein 2/3 complex subunit 1B, 41kDa; actin related protein 2/3 complex, subunit 1B, 41kDa; Actin-related protein 2/3 complex subunit 1B; Arp2/3 complex 41 kDa subunit; ARP2/3 protein complex subunit p41; OTTHUMP00000206020; p41-ARC
Gene Aliases: 41kDa; AA408064; AA408534; AA571392; AF007010; ARC41; ARPC1B; AW208418; L72; p40-ARC; p41-ARC; SOP2Hs
UniProt ID: (Human) O15143, (Mouse) Q9WV32
Entrez Gene ID: (Human) 10095, (Mouse) 11867
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support