Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse ARSA, recombinant human (rh) ARSB and rhARSJ is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Sulfatases, such as ARSG, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules. This gene, a novel arylsulfatase, has activity toward pseudosubstrates including p-nitrocatechol sulfate and 4-methylumbelliferyl sulfate. Activity is competitively inhibited by phosphate. The unprocessed protein is active as a 63-kDa monomer and demonstrates an acidic pH optimum as typically seen for lysosomal sulfatases. The protein accumulates within lysosomes and is also a glycoprotein that binds specifically to mannose 6-phosphate receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arylsulfatase G; ASG; N-sulfoglucosamine-3-sulfatase
Gene Aliases: 6330406P08Rik; AI846872; Arsg; Kiaa1001
UniProt ID: (Mouse) Q3TYD4
Entrez Gene ID: (Mouse) 74008
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support