Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aspartate tRNA ligase 1, cytoplasmic; Aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; AspRS; Cell proliferation-inducing gene 40 protein; testicular tissue protein Li 192
Gene Aliases: 5730439G15Rik; aspRS; DARS; DARS1; HBSL; PIG40
UniProt ID: (Human) P14868, (Mouse) Q922B2
Entrez Gene ID: (Human) 1615, (Mouse) 226414
Molecular Function: aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support