Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Probable disease resistance protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pNd11; Probable disease resistance protein At5g63020
Gene Aliases: AT5G63020; MJH22.8; MJH22_8
UniProt ID: (Arabidopsis) Q8RXS5
Entrez Gene ID: (Arabidopsis) 836422
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support