Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
May play a role in atherogenesis by immobilizing LDL in the atherial wall.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Atherin; SAM domain containing 1; SAM domain-containing protein 1; Sterile alpha motif domain-containing protein 1
Gene Aliases: SAMD1
UniProt ID: (Human) Q6SPF0
Entrez Gene ID: (Human) 90378
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support