Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Serine/threonine-protein kinase component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. Involved in the bipolar attachment of spindle microtubules to kinetochores and is a key regulator for the onset of cytokinesis during mitosis. Required for central/midzone spindle assembly and cleavage furrow formation. Key component of the cytokinesis checkpoint, a process required to delay abscission to prevent both premature resolution of intercellular chromosome bridges and accumulation of DNA damage: phosphorylates CHMP4C, leading to retain abscission-competent VPS4 (VPS4A and/or VPS4B) at the midbody ring until abscission checkpoint signaling is terminated at late cytokinesis (PubMed:22422861, PubMed:24814515). AURKB phosphorylates the CPC complex subunits BIRC5/survivin, CDCA8/borealin and INCENP. Phosphorylation of INCENP leads to increased AURKB activity. Other known AURKB substrates involved in centromeric functions and mitosis are CENPA, DES/desmin, GPAF, KIF2C, NSUN2, RACGAP1, SEPT1, VIM/vimentin, GSG2/Haspin, and histone H3. A positive feedback loop involving GSG2 and AURKB contributes to localization of CPC to centromeres. Phosphorylation of VIM controls vimentin filament segregation in cytokinetic process, whereas histone H3 is phosphorylated at 'Ser-10' and 'Ser-28' during mitosis (H3S10ph and H3S28ph, respectively). A positive feedback between GSG2 and AURKB contributes to CPC localization. AURKB is also required for kinetochore localization of BUB1 and SGOL1. Phosphorylation of p53/TP53 negatively regulates its transcriptional activity. Key regulator of active promoters in resting B- and T-lymphocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIM-1; ARK-1; ARK-2; ARK-3; Aurora 1; Aurora 2; Aurora 3; aurora A; aurora B; aurora family kinase 1; Aurora kinase A; Aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora kinase C; Aurora- and IPL1-like midbody-associated protein 1; aurora-1; aurora-B; aurora-related kinase 1; aurora-related kinase 2; aurora-related kinase 3; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; Aurora/IPL1-related kinase 2; Aurora/IPL1-related kinase 3; Aurora/IPL1/Eg2 protein 1; Aurora/IPL1/Eg2 protein 2; Breast tumor-amplified kinase; epididymis secretory protein Li 90; Ipl1- and aurora-related kinase 1; protein phosphatase 1, regulatory subunit 47; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase 12; serine/threonine kinase 13 (aurora/IPL-like); serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine kinase 5; serine/threonine kinase 6; serine/threonine protein kinase 15; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 13; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase aurora-C; Serine/threonine-protein kinase Ayk1; STK-1
Gene Aliases: AIE1; AIE2; AIK; AIK2; AIK3; AIM-1; AIM1; AIRK1; AIRK2; AIRK3; AL022959; ARK-1; ARK-3; ARK1; ARK2; ARK3; AU019385; AURA; AurB; AurC; AURKA; AURKB; aurkb-sv1; aurkb-sv2; AURKC; Aurora-A; aurora-C; AW539821; AYK1; BTAK; HEL-S-90; IAK; IAK1; IAK3; IPL1; PPP1R47; PPP1R48; SPGF5; STK-1; STK1; STK12; STK13; STK15; STK5; STK6; STK7
UniProt ID: (Human) O14965, (Human) Q96DV5, (Human) Q9UQB9, (Mouse) P97477, (Mouse) O70126, (Mouse) O88445
Entrez Gene ID: (Human) 6790, (Human) 9212, (Human) 6795, (Mouse) 20878, (Mouse) 20877, (Mouse) 20871
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support