Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Recommended positive controls: PC-3, U87-MG, SK-N-SH.
Predicted reactivity: Rhesus Monkey (95%), Bovine (80%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the beta-1, 3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesized by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). This gene is oriented telomere to centromere in close proximity to the ribosomal protein S18 gene. The functionality of the encoded protein is limited to ganglioseries glycolipid biosynthesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: b3Gal-T4; Beta-1,3-galactosyltransferase 4; Beta-1,3-GalTase 4; beta-3-galactosyltransferase 4; Gal-T2; Ganglioside galactosyltransferase; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 4; UDP-galactose:beta-N-acetyl-galactosamine-beta-1,3-galactosyltransferase
Gene Aliases: B3GALT4; BETA3GALT4; GALT2; GALT4
UniProt ID: (Human) O96024
Entrez Gene ID: (Human) 8705
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support