Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLLRRAWRR RAALGLGTLA LCGAALLYLA RCAAEPGDPR AMSGRSPPPP APARAAAFLA VLVASAPRAA ERRSVIRSTW LARRGAPGDV WARFAVGTAG LGAEERRALE REQARHGDLL LLPALRDAYE NLTAKVLAML AWLDEHVAFE FVLKADDDSF ARLDALLAEL RAREPARRRR LYWGFFSGRG RVKPGGRWRE AAWQLCDYYL PYALGGGYVL SADLVHYLRL SRDYLRAWHS EDVSLGAWLA PVDVQREHDP RFDTEYRSRG CSNQYLVTHK QSLEDMLEKH ATLAREGRLC KREVQLRLSY VYDWSAPPSQ CCQRREGIP
B3GALT6, Beta-1, 3-galactosyltransferase that transfers galactose from UDP-galactose to substrates with a terminal beta-linked galactose residue. B3GALT6 has a preference for galactose-beta-1, 4-xylose that is found in the linker region of glycosaminoglycans, such as heparan sulfate and chondroitin sulfate. B3GALT6 has no activity towards substrates with terminal glucosamine or galactosamine residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-galactosyltransferase 6; Beta-1,3-GalTase 6; beta3Gal-T6; GAG GalTII; Galactosyltransferase II; Galactosylxylosylprotein 3-beta-galactosyltransferase; UDP-Gal:betaGal beta 1,3-galactosyltransferase 6; UDP-Gal:betaGal beta 1,3-galactosyltransferase polypeptide 6; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 6
Gene Aliases: B3GALT6; beta3GalT6; EDSP2; SEMDJL1
UniProt ID: (Human) Q96L58
Entrez Gene ID: (Human) 126792
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support