Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
B3GNT5 is a member of the beta-1,3-N-acetylglucosaminyltransferase family. This enzyme is a type II membrane protein. It exhibits strong activity to transfer GlcNAc to glycolipid substrates and is identified as the most likely candidate for lactotriaosylceramide synthase. This enzyme is essential for the expression of Lewis X epitopes on glycolipids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta 1,3 N-acetyglucosaminyltransferase Lc3 synthase; beta-1,3-Gn-T5; beta-1,3-N-acetylglucosaminyltransferase 5; beta-1,3-N-acetylglucosaminyltransferase bGnT-5; beta3Gn-T5; BGnT-5; Lactosylceramide 1,3-N-acetyl-beta-D-glucosaminyltransferase; Lactotriaosylceramide synthase; Lc(3)Cer synthase; lc3 synthase; UDP-GlcNAc:beta-Gal beta-1,3-N-acetylglucosaminyltransferase 5
Gene Aliases: B3GN-T5; B3GNT5; beta3Gn-T5; beta3GnT5
UniProt ID: (Human) Q9BYG0, (Mouse) Q8BGY6
Entrez Gene ID: (Human) 84002, (Mouse) 108105
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support