Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLWKKTVYR SLCLALALLV AVTVFQRSLT PGQFLQEPPP PTLEPQKAQK PNGQLVNPNN FWKNPKDVAA PTPMASQGPQ AWDVTTTNCS ANINLTHQPW FQVLEPQFRQ FLFYRHCRYF PMLLNHPEKC RGDVYLLVVV KSVITQHDRR EAIRQTWGRE RQSAGGGRGA VRTLFLLGTA SKQEERTHYQ QLLAYEDRLY GDILQWGFLD TFFNLTLKEI HFLKWLDIYC PHVPFIFKGD DDVFVNPTNL LEFLADRQPQ ENLFVGDVLQ HARPIRRKDN KYYIPGALYG KASYPPYAGG GGFLMAGSLA RRLHHACDTL ELYPIDDVFL GMCLEVLGVQ PTAHEGFKTF GISRNRNSRM NKEPCFFRAM LVVHKLLPPE LLAMWGLVHS NLTCSRKLQV L
B3GNT7 is a glycosyltransferase which may play a role in elongation of the keratan sulfate carbohydrate backbone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta 1,3-N-acetylglucosaminyltransferase 7; beta-1,3-Gn-T7; beta-1,3-N-acetylglucosaminyltransferase 7; beta3Gn-T7; BGnT-7; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7
Gene Aliases: B3GNT7; beta3GnT7
UniProt ID: (Human) Q8NFL0
Entrez Gene ID: (Human) 93010
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support