Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGSPRAARPP LLLRPVKLLR RRFRLLLALA VVSVGLWTLY LELVASAQVG GNPLNRRYGS WRELAKALAS RNIPAVDPHL QFYHPQRLSL EDHDIDQGVS SNSSYLKWNK PVPWLSEFRG RANLHVFEDW CGSSIQQLRR NLHFPLYPHI RTTLRKLAVS PKWTNYGLRI FGYLHPFTDG KIQFAIAADD NAEFWLSLDD QVSGLQLLAS VGKTGKEWTA PGEFGKFRSQ ISKPVSLSAS HRYYFEVLHK QNEEGTDHVE VAWRRNDPGA KFTIIDSLSL SLFTNETFLQ MDEVGHIPQT AASHVDSSNA LPRDEQPPAD MLRPDPRDTL YRVPLIPKSH LRHVLPDCPY KPSYLVDGLP LQRYQGLRFV HLSFVYPNDY TRLSHMETHN KCFYQENAYY QDRFSFQEYI KIDQPEKQGL EQPGFEENLL EESQYGEVAE ETPASNNQNA RMLEGRQTPA STLEQDATDY RLRSLRKLLA QPREGLLAPF SKRNSTASFP GRTSHIPVQQ PEKRKQKPSP EPSQDSPHSD KWPPGHPVKN LPQMRGPRPR PAGDSPRKTQ WLNQVESYIA EQRRGDRMRP QAPGRGWHGE EEVVAAAGQE GQVEGEEEGE EEEEEEDMSE VFEYVPVFDP VVNWDQTFSA RNLDFQALRT DWIDLSCNTS GNLLLPEQEA LEVTRVFLKK LNQRSRGRYQ LQRIVNVEKR QDQLRGGRYL LELELLEQGQ RVVRLSEYVS ARGWQGIDPA GGEEVEARNL QGLVWDPHNR RRQVLNTRAQ EPKLCWPQGF SWSHRAVVHF VVPVKNQARW VQQFIKDMEN LFQVTGDPHF NIVITDYSSE DMDVEMALKR SKLRSYQYVK LSGNFERSAG LQAGIDLVKD PHSIIFLCDL HIHFPAGVID AIRKHCVEGK MAFAPMVMRL HCGATPQWPE GYWEVNGFGL LGIYKSDLDR IGGMNTKEFR DRWGGEDWEL LDRILQAGLD VERLSLRNFF HHFHSKRGMW SRRQMKTL
Transfers N-acetylgalactosamine (GalNAc) from UDP-GalNAc to N-acetylglucosamine-beta-benzyl with a beta-1,4-linkage to form N,N'-diacetyllactosediamine, GalNAc-beta-1,4-GlcNAc structures in N-linked glycans and probably O-linked glycans. Mediates the N,N'- diacetyllactosediamine formation on gastric mucosa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B4GalNAcT3; beta-1,4-N-acetyl-galactosaminyl transferase 3; Beta-1,4-N-acetylgalactosaminyltransferase 3; Beta-1,4-N-acetylgalactosaminyltransferase III; Beta4GalNAc-T3; beta4GalNAcT3; N-acetyl-beta-glucosaminyl-glycoprotein 4-beta-N- acetylgalactosaminyltransferase; N-acetyl-beta-glucosaminyl-glycoprotein 4-beta-N-acetylgalactosaminyltransferase 2; NGalNAc-T2
Gene Aliases: B4GALNT3
UniProt ID: (Human) Q6L9W6
Entrez Gene ID: (Human) 283358
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support