Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
Description: This MA57YW monoclonal antibody recognizes human B7-H7 on live and paraformaldehyde-fixed cells.
Applications Reported: This MA57YW antibody has been reported for use in flow cytometric analysis. This MA57YW monoclonal antibody recognizes human B7-H7 on live and paraformaldehyde-fixed cells.
Applications Tested: This MA57YW antibody has been pre-diluted and tested by flow cytometric analysis of the HDLM-2 cell line. This may be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: HERV-H LTR-associating protein 2; Human endogenous retrovirus-H long terminal repeat-associating protein 2
Gene Aliases: B7-H5; B7-H7; B7H7; B7y; HHLA2
UniProt ID: (Human) Q9UM44
Entrez Gene ID: (Human) 11148
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support