Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPNDSTSTA VEEPDSLEVL VKTLDSQTRT FIVGAQMNVK EFKEHIAASV SIPSEKQRLI YQGRVLQDDK KLQEYNVGGK VIHLVERAPP QTHLPSGASS GTGSASATHG GGSPPGTRGP GASVHDRNAN SYVMVGTFNL PSDGSAVDVH INMEQAPIQS EPRVRLVMAQ HMIRDIQTLL SRMECRGGPQ PQHSQPPPQP PAVTPEPVAL SSQTSEPVES EAPPREPMEA EEVEERAPAQ NPELTPGPAP AGPTPAPETN APNHPSPAEY VEVLQELQRL ESRLQPFLQR YYEVLGAAAT TDYNNNHEGR EEDQRLINLV GESLRLLGNT FVALSDLRCN LACTPPRHLH VVRPMSHYTT PMVLQQAAIP IQINVGTTVT MTGNGTRPPP TPNAEAPPPG PGQASSVAPS STNVESSAEG APPPGPAPPP ATSHPRVIRI SHQSVEPVVM MHMNIQDSGT QPGGVPSAPT GPLGPPGHGQ TLGQQVPGFP TAPTRVVIAR PTPPQARPSH PGGPPVSGTL QGAGLGTNAS LAQMVSGLVG QLLMQPVLVA QGTPGMAPPP APATASASAG TTNTATTAGP APGGPAQPPP TPQPSMADLQ FSQLLGNLLG PAGPGAGGSG VASPTITVAM PGVPAFLQGM TDFLQATQTA PPPPPPPPPP PPAPEQQTMP PPGSPSGGAG SPGGLGLESL SPEFFTSVVQ GVLSSLLGSL GARAGSSESI AAFIQRLSGS SNIFEPGADG ALGFFGALLS LLCQNFSMVD VVMLLHGHFQ PLQRLQPQLR SFFHQHYLGG QEPTPSNIRM ATHTLITGLE EYVRESFSLV QVQPGVDIIR TNLEFLQEQF NSIAAHVLHC TDSGFGARLL ELCNQGLFEC LALNLHCLGG QQMELAAVIN GRIRRMSRGV NPSLVSWLTT MMGLRLQVVL EHMPVGPDAI LRYVRRVGDP PQPLPEEPME VQGAERASPE PQRENASPAP GTTAEEAMSR GPPPAPEGGS RDEQDGASAE TEPWAAAVPP EWVPIIQQDI QSQRKVKPQP PLSDAYLSGM PAKRRKTMQG EGPQLLLSEA VSRAAKAAGA RPLTSPESLS RDLEAPEVQE SYRQQLRSDI QKRLQEDPNY SPQRFPNAQR AFADDP
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAG 6; BAG family molecular chaperone regulator 6; BAG-6; BAT 3; BCL2-associated athanogene 6; DAQB-195H10.3; HLA-B associated transcript 3; HLA-B associated transcript-3; HLA-B-associated transcript 3; Large proline-rich protein BAG6; large proline-rich protein BAT3; Protein G3; Protein Scythe; scythe
Gene Aliases: 2410045D21Rik; AA408914; BAG-6; BAG6; BAT3; D17H6S52E; D6S52E; G3; Scythe
UniProt ID: (Human) P46379, (Rat) Q6MG49, (Mouse) Q9Z1R2
Entrez Gene ID: (Human) 7917, (Rat) 94342, (Mouse) 224727
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support