Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
BATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events and blocks cellular transformation by Ras and Fos. BATF also is required for the differentiation of IL-17-producing T helper (TH17) cells, a CD4+ subset of T cells that coordinates the inflammatory response in host defense. In both T cells and B cells, BATF is required for the appropriate regulation of the class-switch recombination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activating transcription factor B; B-ATF; B-cell activating transcription factor; B-cell-activating transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; Basic leucine zipper transcriptional factor ATF-like; SF-HT-activated gene 2 protein; SFA-2
Gene Aliases: B-ATF; BATF; BATF1; SFA-2; SFA2
UniProt ID: (Human) Q16520, (Mouse) O35284
Entrez Gene ID: (Human) 10538, (Mouse) 53314
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support