Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: GNKDLKKRKM EENTSINLSK QESFTSVKKP KRDDSKDLAL CSMILTEMET HEDAWPFLLP VNLKLVPGYK KVIKKPMDFS TIREKLSSGQ YPNLETFAL
This gene belongs to the bromodomain gene family. Members of this gene family encode proteins that are integral components of chromatin remodeling complexes. The encoded protein showed strong preference for the activating H3K14Ac mark in a histone peptide screen, suggesting a potential role in transcriptional activation. This gene may be associated with susceptibility to sudden cardiac death (SCD). [provided by RefSeq, Aug 2016].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bromodomain adjacent to zinc finger domain protein 2B; bromodomain adjacent to zinc finger domain, 2B; DKFZP434H071; DKFZp762I0516; FLJ45644; hWALp4
Gene Aliases: BAZ2B; KIAA1476; WALp4
UniProt ID: (Human) Q9UIF8
Entrez Gene ID: (Human) 29994
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support