Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Probable molecular chaperone. Assists the folding of proteins upon ATP hydrolysis. As part of the BBS/CCT complex may play a role in the assembly of BBSome, a complex involved in ciliogenesis regulating transports vesicles to the cilia. May play a role in protein processing in limb, cardiac and reproductive system development. May play a role in cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bardet-Biedl syndrome 6 protein; McKusick-Kaufman syndrome protein; McKusick-Kaufman syndrome protein; MKKS protein; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; Molecular chaperone MKKS; Protein Bbs6 homolog
Gene Aliases: 1300013E18Rik; AI463362; AI957237; BBS6; HMCS; KMS; MKKS; MKS
UniProt ID: (Human) Q9NPJ1, (Mouse) Q9JI70
Entrez Gene ID: (Human) 8195, (Mouse) 59030, (Rat) 311456
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support