Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Promo Code: CorderMand2 .
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MTQKPHKKPG PGEPLVFGKT FTDHMLMVEW NDKGWGQPRI QPFQNLTLHP ASSSLHYSLQ LFEGMKAFKG KDQQVRLFRP WLNMDRMLRS AMRLCLPSFD KLELLECIRR LIEVDKDWVP DAAGTSLYVR PVLIGNEPSL GVSQPTRALL FVILCPVGAY FPGGSVTPVS LLADPAFIRA WVGGVGNYKL GGNYGPTVLV QQEALKRGCE QVLWLYGPDH QLTEVGTMNI FVYWTHEDGV LELVTPPLNG VILPGVVRQS LLDMAQTWGE FRVVERTITM KQLLRALEEG RVREVFGSGT ACQVCPVHRI LYKDRNLHIP TMENGPELIL RFQKELKEIQ YGIRAHEWMF PV
This gene encodes a branched chain aminotransferase found in mitochondria. The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine, isoleucine, and valine. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCAT(m); branched chain amino-acid transaminase 2, mitochondrial; branched chain aminotransferase 2, mitochondrial; Branched-chain-amino-acid aminotransferase, mitochondrial; Placental protein 18; PP18
Gene Aliases: BCAM; Bcat-2; BCAT2; BCATM; BCT2; ECA40; PP18
UniProt ID: (Human) O15382, (Mouse) O35855
Entrez Gene ID: (Human) 587, (Mouse) 12036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support