Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This is a rabbit polyclonal antibody raised against part chain of N-terminal BCL11A of human origin. There are three isoforms of BCL11A: isoform1 (773 aa), isoform2 (486 aa), and isoform3 (239 aa).
Immunogen sequence: MSRRKQGKP QHLSKREFSP EPLEAILTDD EPDHGPLGAP EGDHDLLTCG QCQMNFPLGD ILIFIEHKRK QCNGSLCLEK AVDKPPSPSP IEMKKASNPV EVGIQVTPED DDCLSTSSRG ICPKQEHIAD KLLHWRGLSS PRSAHGALIP TPGMSAEYAP QGICKDEPSS YTCTTCKQPF TSAWFLLQHA QNTHGLRIYL ESEHGSPLTP RVGIPSGLGA ECPSQPPLHG IHIADNNPFN LLRIPGSVSR EASGLAEGRF PPTPPLFSPP PRHHLDPHRI ERLGAEEMAL ATHHPSAFDR VLRLNPMAME PPAMDFSRRL RELAGNTSSP PLSPGRPSPM QRLLQPFQPG SK (1-351 aa encoded by BC021098)
This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell CLL/lymphoma 11A; B-cell CLL/lymphoma 11A (zinc finger protein); B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2; B-cell lymphoma/leukemia 11A; BCL-11A; BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1; C2H2-type zinc finger protein; COUP-TF interacting protein 1; COUP-TF-interacting protein 1; ecotropic viral integration site 9 homolog; Ecotropic viral integration site 9 protein; Ecotropic viral integration site 9 protein homolog; EVI-9; FLJ10173; FLJ34997; myeloid leukemia; Zinc finger protein 856
Gene Aliases: 2810047E18Rik; BCL-11A; BCL11A; BCL11A-L; BCL11a-M; BCL11A-S; BCL11A-XL; CTIP1; D930021L15Rik; EVI9; Evi9a; Evi9b; Evi9c; HBFQTL5; KIAA1809; mKIAA1809; ZNF856
UniProt ID: (Human) Q66LN6, (Mouse) Q9QYE3
Entrez Gene ID: (Human) 53335, (Rat) 305589, (Mouse) 14025
Molecular Function:
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support