Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NLPTPLYPM MCPMEHPLSA DIAMATRADE DGDTPLHIAV VQGNLPAVHR LVNLFQQGGR ELDIYNNLRQ TPLHLAVITT LPSVVRLLVT AGASPMALDR HGQTAAHLAC EHRSPTCLRA LLDSAAPGTL DLEARNYDGL TALHVAVNTE CQETVQLLLE RGADIDAVDI KSGRSPLIHA VENNSLSMVQ LLLQHGANVN AQMYSGSSAL HSASGRGLLP LVRTLVRSGA DSSLKNCHND TPLMVARSRR VIDILRGKAT RPASTSQPDP SPDRSANTSP ESSSRLSSNG LLSASPSSSP SQSPPRDPPG FPMAPPNFFL PSPSPPAFLP FAGVLRGPGR PVPPSPAPGG S (97-446 aa encoded by BC064993)
B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL-3; Bcl-3 protein; chronic lymphatic leukemia protein; Proto-oncogene BCL3
Gene Aliases: AI528691; Bcl-3; BCL3; BCL4; D19S37
UniProt ID: (Human) P20749, (Mouse) Q9Z2F6
Entrez Gene ID: (Human) 602, (Mouse) 12051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support