Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MERGMHLGA AAAGEDDLFL HKSLSASTSK RLEAAFRSTP PGMDLSLAPP PRERPASSSS SPLGCFEPAD PEGAGLLLPP PGGGGGGSAG SGGGGGGGVG VPGLLVGSAG VGGDPSLSSL PAGAALCLKY GESASRGSVA ESSGGEQSPD DDSDGRCELV LRAGVADPRA SPGAGGGGAK AAEGCSNAHL HGGASVPPGG LGGGGGGGSS SGSSGGGGGS GSGSGG (1-225 aa encoded by B C156671)
BHLHB5 inhibits DNA binding of TCF3/E47 homodimers and TCF3 (E47)/NEUROD1 heterodimers and acts as a strong repressor of Neurod1 and Myod-responsive genes, probably by heterodimerization with class a basic helix-loop-helix factors. BHLHB5 despite the presence of an intact basic domain, does not bind to DNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic helix-loop-helix (bHLH) gene, class B, beta3; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix domain containing, class B5; basic helix-loop-helix family, member e22; bHLHb5; bHLHe22; Class B basic helix-loop-helix protein 5; Class E basic helix-loop-helix protein 22; Protein BETA3; trinucleotide repeat containing 20; Trinucleotide repeat-containing gene 20 protein
Gene Aliases: Beta3; BHLHB5; BHLHE22; CAGL85; TNRC20
UniProt ID: (Human) Q8NFJ8, (Mouse) Q8C6A8
Entrez Gene ID: (Human) 27319, (Mouse) 59058
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support