Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • BHLHE40 Antibodies

            OriGene

            BHLHE40 Monoclonal Antibody (OTI1F10), TrueMAB™

            View all (22) BHLHE40 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # CF810131
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            662.00
            100 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite BHLHE40 Monoclonal Antibody (OTI1F10), TrueMAB™

            • Antibody Testing Data (1)
            Application
            BHLHE40 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            BHLHE40 Antibody (CF810131) in WB

            HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY BHLHE40 (RC210294, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-BHLH. Positive lysates LY418509 (100 µg) and LC418509 (20 µg) can be purchased separately from OriGene.
            View Product

            BHLHE40 Antibody in Western Blot (WB)

            Product Details

            CF810131

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Mouse / IgG1

            Class

            Monoclonal

            Type

            Antibody

            Clone

            OTI1F10

            Immunogen

            Human recombinant protein fragment corresponding to amino acids 1-313 of human BHLHE40 produced in E.coli.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Concentration

            1 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 8% trehalose

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

            Target Information

            Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: 1-Dec; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family, member e40; bHLHb2; bHLHe40; Class B basic helix-loop-helix protein 2; Class E basic helix-loop-helix protein 40; cytokine response gene 8; DEC1; differentially expressed in chondrocytes 1; Differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of split and hairy-related protein 2; Enhancer-of-split and hairy-related protein 2; FLJ99214; SHARP-2; stimulated by retinoic acid 14; Stimulated by retinoic acid gene 13 protein

            View more View less

            Gene Aliases: BHLHB2; BHLHE40; C130042M06Rik; Clast5; CR8; DEC1; HLHB2; SHARP-2; SHARP2; STRA13; Stra14

            View more View less

            UniProt ID: (Human) O14503, (Mouse) O35185, (Rat) O35780

            View more View less

            Entrez Gene ID: (Human) 8553, (Mouse) 20893, (Rat) 79431

            View more View less

            Function(s)

            RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II activating transcription factor binding transcriptional repressor activity, RNA polymerase II transcription factor binding transcription factor activity, sequence-specific DNA binding transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding transcription corepressor activity protein binding protein domain specific binding protein homodimerization activity bHLH transcription factor binding MRF binding protein heterodimerization activity E-box binding DNA binding protein dimerization activity basic helix-loop-helix transcription factor

            Process(es)

            negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated circadian regulation of gene expression entrainment of circadian clock by photoperiod negative regulation of sequence-specific DNA binding transcription factor activity negative regulation of transcription, DNA-templated transcription from RNA polymerase II promoter response to light stimulus entrainment of circadian clock rhythmic process nervous system development circadian rhythm regulation of neuronal synaptic plasticity
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-6jbw8:80/100.66.73.217:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0