Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total BHLHB3.
BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-Dec; basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix domain containing, class B3; basic helix-loop-helix family, member e41; bHLH transcriptional factor Dec2; bHLHb3; bHLHe41; Class B basic helix-loop-helix protein 3; Class E basic helix-loop-helix protein 41; Differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; mDEC2; SHARP-1
Gene Aliases: 6430520M22Rik; Bhlhb2l; BHLHB3; BHLHE41; DEC2; hDEC2; SHARP-1; SHARP1
UniProt ID: (Human) Q9C0J9, (Mouse) Q99PV5
Entrez Gene ID: (Human) 79365, (Rat) 117095, (Mouse) 79362
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support