Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAPAGRPGAK KGILERLESG EVVIGDGSFL ITLEKRGYVK AGLWTPEAVI EHPDAVRQLH MEFLRAGSNV MQTFTFSASE DNMESKWEDV NAAACDLARE VAGKGDALVA GGICQTSIYK YQKDEARIKK LFRQQLEVFA WKNVDFLIAE YFEHVEEAVW AVEVLKESDR PVAVTMCIGP EGDMHDITPG ECAVRLVKAG ASIVGVNCRF GPDTSLKTME LMKEGLEWAG LKAHLMVQPL GFHAPDCGKE GFVDLPEYPF GLESRVATRW DIQKYAREAY NLGVRYIGGC CGFEPYHIRA IAEELAPERG FLPPASEKHG SWGSGLDMHT KPWIRARARR EYWENLLPAS GRPFCPSLSK PDF
Homocysteine is a sulfur-containing amino acid that plays a crucial role in methylation reactions. Transfer of the methyl group from betaine to homocysteine creates methionine, which donates the methyl group to methylate DNA, proteins, lipids, and other intracellular metabolites. The protein encoded by this gene is one of two methyl transferases that can catalyze the transfer of the methyl group from betaine to homocysteine. Anomalies in homocysteine metabolism have been implicated in disorders ranging from vascular disease to neural tube birth defects such as spina bifida.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Betaine--homocysteine S-methyltransferase 2; betaine-homocysteine methyltransferase 2; S-methylmethionine--homocysteine S-methyltransferase BHMT2; SMM-hcy methyltransferase
Gene Aliases: BHMT2
UniProt ID: (Human) Q9H2M3
Entrez Gene ID: (Human) 23743
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support