Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor guanine nucleotide-exchange factor 1; ADP-ribosylation factor guanine nucleotide-exchange factor 1 (brefeldin A-inhibited); BIG1; Brefeldin A-inhibited GEP 1; Brefeldin A-inhibited guanine nucleotide-exchange protein 1; DKFZP434L057; p200 ARF guanine nucleotide exchange factor; p200 ARF-GEP1
Gene Aliases: ARFGEF1; ARFGEP1; BIG1; D130059B05Rik; D730028O18Rik; P200; RGD1560793
UniProt ID: (Human) Q9Y6D6, (Rat) D4A631, (Mouse) G3X9K3
Entrez Gene ID: (Human) 10565, (Rat) 312915, (Mouse) 211673
Molecular Function:
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support